Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173452.1 | NP_001166923.1 | 14 | 2166 | ||
NM_001173453.1 | NP_001166924.1 | 13 | 2013 | |||
NM_005653.4 | NP_005644.2 | 14 | 2166 | |||
GenBank mRNA | AK291264.1 | - | 14 | 1817 | ||
BC003634.2 | - | 14 | 1904 | 3529119 | ||
M84810.1 | - | 14 | 1808 | |||
U03494.1 | - | 14 | 2136 | |||
U03495.1 | - | 14 | 1800 |