Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003810.1 | NP_001003810.1 | 7 | 1640 | ||
NM_002138.3 | NP_002129.2 | 8 | 1697 | |||
NM_031369.2 | NP_112737.1 | 8 | 1787 | |||
NM_031370.2 | NP_112738.1 | 9 | 1844 | |||
GenBank mRNA | AF039575.1 | - | 8 | 1355 | ||
BG180941.1 | - | 1 | 183 |