Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313726.1 | NP_001300655.1 | 4 | 497 | ||
NM_031418.3 | NP_113606.2 | 3 | 405 | |||
GenBank mRNA | AJ300461.1 | - | 3 | 1112 | ||
AK302659.1 | - | 3 | 299 | |||
AK304781.1 | - | 4 | 338 | |||
DA247850.1 | - | 3 | 365 |