Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031438.3 | NP_113626.1 | 2 | 277 | ||
XM_005272095.1 | XP_005272152.1 | 2 | 262 | |||
GenBank mRNA | AK098066.1 | - | 2 | 246 | ||
AK309942.1 | - | 2 | 262 | |||
AL136592.1 | - | 2 | 232 | |||
BC041099.1 | - | 2 | 261 | 5495144 | ||
CB131118.1 | - | 2 | 265 | |||
DB029218.1 | - | 2 | 279 |