Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISPLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031476.3 | NP_113664.1 | 2 | 403 | ||
XM_005256190.1 | XP_005256247.1 | 3 | 658 | |||
GenBank mRNA | AK027395.1 | - | 2 | 367 | ||
AK303887.1 | - | 2 | 399 | |||
AL136861.1 | - | 2 | 380 | |||
AY358413.1 | - | 2 | 387 | |||
AY358790.1 | - | 2 | 412 | |||
BC007689.2 | - | 2 | 379 | |||
BC063012.1 | - | 2 | 381 | 5217820 | ||
DA325436.1 | - | 2 | 399 | |||
DB095807.1 | - | 2 | 403 |