Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300948.1 | NP_001287877.1 | 6 | 1291 | ||
NM_031490.3 | NP_113678.2 | 7 | 1423 | |||
XM_017023755.1 | XP_016879244.1 | 7 | 1290 | |||
XM_017023756.1 | XP_016879245.1 | 6 | 1156 | |||
GenBank mRNA | AJ548761.1 | - | 7 | 1290 | ||
AK027544.1 | - | 7 | 1256 | |||
AK027666.1 | - | 3 | 458 | |||
AK074775.1 | - | 7 | 1288 | |||
AK127125.1 | - | 7 | 1055 | |||
AK294779.1 | - | 6 | 1103 | |||
AL834201.1 | - | 6 | 789 | |||
BC093910.1 | - | 7 | 1257 | 7939755 | ||
BC093912.1 | - | 7 | 1257 | 7939757 | ||
BC110434.1 | - | 3 | 439 | |||
BC143246.1 | - | 6 | 1125 | 9051747 |