Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031898.2 | NP_114104.1 | 3 | 497 | ||
XM_011523988.2 | XP_011522290.1 | 3 | 392 | |||
XM_011523989.1 | XP_011522291.1 | 2 | 454 | |||
XM_011523990.2 | XP_011522292.1 | 4 | 1814 | |||
XM_017024954.1 | XP_016880443.1 | 3 | 541 | |||
XM_017024955.1 | XP_016880444.1 | 3 | 237 | |||
XM_017024956.1 | XP_016880445.1 | 3 | 497 | |||
GenBank mRNA | AF334676.1 | - | 2 | 409 | ||
AK057390.1 | - | 3 | 497 | |||
AK223534.1 | - | 3 | 453 | |||
AK314327.1 | - | 3 | 489 | |||
BC031688.1 | - | 3 | 460 | 5166920 |