Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321995.1 | NP_001308924.1 | 4 | 402 | ||
NM_001321996.1 | NP_001308925.1 | 4 | 581 | |||
NM_001321997.1 | NP_001308926.1 | 4 | 402 | |||
NM_031902.4 | NP_114108.1 | 4 | 581 | |||
NR_136152.1 | 4 | 581 | ||||
NR_136153.1 | 4 | 581 | ||||
GenBank mRNA | AB049940.1 | - | 4 | 462 | ||
AK058160.1 | - | 4 | 355 | |||
AK295812.1 | - | 4 | 367 | |||
BC014172.2 | - | 4 | 554 | 4561399 | ||
BG771419.1 | - | 4 | 568 |