Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014764.2 | NP_001014764.1 | 2 | 722 | ||
NM_031298.3 | NP_112588.1 | 2 | 609 | |||
GenBank mRNA | BC001409.1 | - | 2 | 517 | 3139577 | |
BM825446.1 | - | 2 | 503 | |||
BQ277705.1 | - | 2 | 623 | |||
BU931404.1 | - | 2 | 623 |