Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042353.2 | NP_001035812.1 | 2 | 1487 | ||
NM_001289145.1 | NP_001276074.1 | 3 | 1596 | |||
NM_001289146.1 | NP_001276075.1 | 2 | 1484 | |||
NM_001289147.1 | NP_001276076.1 | 2 | 1274 | |||
NM_031424.5 | NP_113612.1 | 1 | 1332 | |||
NM_207121.4 | NP_997004.1 | 2 | 1279 | |||
XM_011529381.2 | XP_011527683.1 | 2 | 2020 | |||
GenBank mRNA | BC004222.2 | - | 2 | 1279 | 3532660 | |
BC012800.2 | - | 1 | 1295 | 2961135 | ||
BC022238.1 | - | 1 | 1281 | 4181303 |