Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182663.3 | NP_872604.1 | 6 | 1672 | ||
NM_182664.3 | NP_872605.1 | 5 | 1556 | |||
NM_182665.3 | NP_872606.1 | 5 | 1550 | |||
GenBank mRNA | AB209145.1 | - | 4 | 3632 | ||
AF445801.1 | - | 5 | 1550 | |||
AK056568.1 | - | 2 | 1316 | |||
AK074115.1 | - | 3 | 2547 | |||
AL832784.1 | - | 5 | 1450 | |||
AY062002.1 | - | 6 | 1656 | |||
AY062003.1 | - | 5 | 1540 | |||
AY216268.1 | - | 5 | 1543 | |||
BC004270.1 | - | 7 | 1526 | 3613993 | ||
BC007203.1 | - | 7 | 1526 | 2967057 | ||
BC042651.1 | - | 5 | 1530 | 4342962 |