Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031474.2 | NP_113662.1 | 6 | 1522 | ||
XM_005253797.3 | XP_005253854.1 | 7 | 1534 | |||
XM_005253798.3 | XP_005253855.1 | 6 | 1430 | |||
XM_011521031.2 | XP_011519333.1 | 7 | 1464 | |||
XM_017020000.1 | XP_016875489.1 | 4 | 2095 | |||
XM_017020001.1 | XP_016875490.1 | 7 | 1221 | |||
GenBank mRNA | AK054740.1 | - | 2 | 733 | ||
AK098063.1 | - | 6 | 1493 | |||
AL136557.1 | - | 6 | 1490 | |||
BC036063.1 | - | 6 | 1491 | 5276999 |