Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031484.3 | NP_113672.1 | 2 | 3096 | ||
GenBank mRNA | AF325259.1 | - | 2 | 3083 | ||
AK025996.1 | - | 2 | 2781 | |||
BC004995.1 | - | 2 | 3080 | |||
BC062544.1 | - | 2 | 3082 |