Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300948.1 | NP_001287877.1 | 14 | 2549 | ||
NM_031490.3 | NP_113678.2 | 15 | 2681 | |||
GenBank mRNA | AJ548761.1 | - | 15 | 2548 | ||
AK027544.1 | - | 15 | 2514 | |||
AK027666.1 | - | 11 | 1716 | |||
AK074775.1 | - | 15 | 2546 | |||
AK090452.1 | - | 2 | 3725 | |||
AK127125.1 | - | 15 | 2313 | |||
AK130091.1 | - | 4 | 558 | |||
AK172790.1 | - | 6 | 836 | |||
AL834201.1 | - | 14 | 2047 | |||
BC002916.2 | - | 3 | 517 | |||
BC004234.2 | - | 3 | 517 | |||
BC093910.1 | - | 15 | 2515 | 7939755 | ||
BC093912.1 | - | 15 | 2515 | 7939757 | ||
BC110434.1 | - | 11 | 1697 | |||
BC143246.1 | - | 14 | 2383 | 9051747 |