Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMCHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003921.1 | 4 | 494 | |||
GenBank mRNA | AY008407.1 | - | 5 | 572 | ||
AY008408.1 | - | 5 | 576 | |||
AY008409.1 | - | 4 | 494 | |||
AY008410.1 | - | 4 | 532 | |||
AY008411.1 | - | 4 | 1753 | |||
EU921424.1 | - | 5 | 408 | |||
EU921426.1 | - | 5 | 479 | |||
EU921427.1 | - | 5 | 519 | |||
EU921428.1 | - | 4 | 402 | |||
EU921429.1 | - | 5 | 479 | |||
EU921430.1 | - | 6 | 675 |