Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142624.2 | NP_001136096.2 | 3, 4 | 316 | ||
NM_001144942.1 | NP_001138414.1 | 2, 3 | 768 | |||
NM_001144943.1 | NP_001138415.1 | 3, 4 | 316 | |||
NM_001256277.1 | NP_001243206.1 | 3, 4 | 450 | |||
NM_031934.5 | NP_114140.4 | 2, 3 | 768 | |||
GenBank mRNA | AF322067.1 | - | 2, 3 | 144 | ||
AJ277106.1 | - | 3, 4 | 355 | |||
AK027312.1 | - | 3, 4 | 349 | |||
AK074689.1 | - | 2, 3 | 746 | |||
AK297850.1 | - | 3, 4 | 224 | |||
BC016841.1 | - | 2, 3 | 626 | 3902651 | ||
BC066904.1 | - | 3, 4 | 365 | 5260472 | ||
BC091510.1 | - | 2, 3 | 310 | 30512024 | ||
BT006702.1 | - | 2, 3 | 144 | |||
CB136913.1 | - | 3, 4 | 415 | |||
CX788239.1 | - | 3, 4 | 209 |