Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031948.4 | NP_114154.1 | 3 | 488 | ||
GenBank mRNA | AB056161.1 | - | 3 | 147 | ||
AJ306593.1 | - | 3 | 116 | |||
AK126510.1 | - | 1 | 2668 | |||
AY030095.1 | - | 3 | 97 | |||
AY359106.1 | - | 3 | 132 | |||
BC034294.2 | - | 3 | 152 | 4752547 |