Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031949.4 | NP_114155.4 | 2 | 233 | ||
GenBank mRNA | AK093039.1 | - | 2 | 233 | ||
AK225984.1 | - | 2 | 233 | |||
AK314450.1 | - | 2 | 210 | |||
AY026506.1 | - | 2 | 205 | |||
BC030650.1 | - | 2 | 223 | 5269216 | ||
BC047411.1 | - | 2 | 211 | 5296230 |