Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031952.3 | NP_114158.2 | 3 | 349 | ||
NR_125327.1 | 3 | 349 | ||||
NR_125328.1 | 3 | 324 | ||||
NR_125329.1 | 3 | 349 | ||||
XM_011543664.1 | XP_011541966.1 | 3 | 305 | |||
XM_017009948.1 | XP_016865437.1 | 3 | 330 | |||
XM_017009949.1 | XP_016865438.1 | 8 | 886 | |||
XM_017009950.1 | XP_016865439.1 | 7 | 868 | |||
GenBank mRNA | AK292338.1 | - | 3 | 330 | ||
AK310260.1 | - | 3 | 305 | |||
AY027526.1 | - | 3 | 298 | |||
BC032832.1 | - | 3 | 354 | 4827641 | ||
BC047333.1 | - | 3 | 335 | 5166271 |