Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031955.5 | NP_114161.3 | 2 | 349 | ||
XM_006713778.2 | XP_006713841.1 | 2 | 323 | |||
XM_017007308.1 | XP_016862797.1 | 2 | 323 | |||
GenBank mRNA | AF345909.2 | - | 2 | 295 | ||
AF404758.1 | - | 2 | 295 | |||
AK093041.1 | - | 2 | 297 | |||
BC034496.2 | - | 2 | 251 | 4825905 | ||
BI829426.1 | - | 2 | 349 | |||
DB040560.1 | - | 2 | 323 |