Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 4 | 859 | ||
XM_011518446.2 | XP_011516748.1 | 4 | 877 | |||
XM_011518447.1 | XP_011516749.1 | 4 | 859 | |||
XM_011518451.1 | XP_011516753.1 | 4 | 859 | |||
XM_011518452.1 | XP_011516754.1 | 4 | 859 | |||
XM_017014571.1 | XP_016870060.1 | 4 | 859 | |||
XM_017014572.1 | XP_016870061.1 | 4 | 859 | |||
GenBank mRNA | AF153415.1 | - | 4 | 859 | ||
AK299952.1 | - | 4 | 841 | |||
AL833960.1 | - | 3 | 166 | |||
BP351493.1 | - | 4 | 380 |