Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031947.3 | NP_114153.1 | 1 | 636 | ||
GenBank mRNA | AF332005.1 | - | 1 | 438 | ||
AF378119.1 | - | 1 | 618 | |||
AY208943.1 | - | 1 | 546 | |||
BC069458.1 | - | 1 | 438 | 7262278 | ||
BC100935.2 | - | 1 | 438 | 40004045 | ||
BC100936.1 | - | 1 | 438 | 40004046 | ||
BC100937.2 | - | 1 | 438 | 40004047 |