Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318395.1 | NP_001305324.1 | 5 | 1014 | ||
NM_031948.4 | NP_114154.1 | 6 | 1177 | |||
XM_011522689.2 | XP_011520991.1 | 4 | 906 | |||
GenBank mRNA | AB056161.1 | - | 6 | 836 | ||
AJ306593.1 | - | 6 | 805 | |||
AK055576.1 | - | 3 | 1778 | |||
AY030095.1 | - | 6 | 786 | |||
AY359106.1 | - | 6 | 821 | |||
BC034294.2 | - | 6 | 841 | 4752547 | ||
BC036912.2 | - | 5 | 1025 |