Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317781.1 | NP_001304710.1 | 9 | 1454 | ||
XM_005263267.4 | XP_005263324.1 | 9 | 1897 | |||
XM_011532302.2 | XP_011530604.1 | 11 | 2032 | |||
XM_011532303.2 | XP_011530605.1 | 11 | 2029 | |||
XM_011532304.2 | XP_011530606.1 | 10 | 1996 | |||
XM_011532305.2 | XP_011530607.1 | 10 | 1933 | |||
XM_011532306.2 | XP_011530608.1 | 11 | 2032 | |||
XM_011532307.2 | XP_011530609.1 | 11 | 2032 | |||
XM_011532308.2 | XP_011530610.1 | 10 | 1996 | |||
XM_017008680.1 | XP_016864169.1 | 10 | 1996 | |||
XM_017008681.1 | XP_016864170.1 | 11 | 2032 | |||
XM_017008682.1 | XP_016864171.1 | 9 | 1897 | |||
XM_017008683.1 | XP_016864172.1 | 8 | 941 | |||
XM_017008684.1 | XP_016864173.1 | 10 | 1213 | |||
XM_017008685.1 | XP_016864174.1 | 4 | 659 | |||
GenBank mRNA | AF113223.1 | - | 3 | 339 | ||
AK026814.1 | - | 2 | 196 | |||
AY044656.1 | - | 2 | 254 |