Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320823.1 | NP_001307752.1 | 6 | 531 | ||
NM_001320824.1 | NP_001307753.1 | 6 | 586 | |||
NM_001320825.1 | NP_001307754.1 | 5 | 330 | |||
NM_032118.3 | NP_115494.1 | 6 | 553 | |||
XM_017005064.1 | XP_016860553.1 | 5 | 332 | |||
GenBank mRNA | AK023015.1 | - | 6 | 546 | ||
AK222696.1 | - | 6 | 546 | |||
AK311621.1 | - | 2 | 233 | |||
BC051848.1 | - | 6 | 518 | 6150670 | ||
BG024606.1 | - | 6 | 552 | |||
BI855898.1 | - | 6 | 494 | |||
CD249537.1 | - | 5 | 346 |