Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184957.1 | NP_001171886.1 | 4 | 513 | ||
NM_001184958.1 | NP_001171887.1 | 4 | 618 | |||
NM_032137.4 | NP_115513.4 | 4 | 1015 | |||
XM_006713341.2 | XP_006713404.1 | 4 | 1015 | |||
XM_006713342.2 | XP_006713405.1 | 4 | 868 | |||
XM_006713343.2 | XP_006713406.1 | 4 | 1015 | |||
XM_011534153.2 | XP_011532455.1 | 4 | 2645 | |||
XM_011534154.2 | XP_011532456.1 | 4 | 2080 | |||
XM_011534158.2 | XP_011532460.1 | 4 | 1015 | |||
XM_017007310.1 | XP_016862799.1 | 4 | 1015 | |||
GenBank mRNA | AK131481.1 | - | 4 | 869 | ||
AL136781.1 | - | 4 | 433 | |||
AL834386.1 | - | 4 | 967 | |||
BC030599.2 | - | 4 | 576 | 4831210 | ||
BC038406.1 | - | 4 | 935 | 5167835 | ||
BG718869.1 | - | 4 | 581 |