Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091344.1 | - | 1 | 1956 | ||
AK299614.1 | - | 3 | 1590 | |||
AK315589.1 | - | 1 | 1951 | |||
AL136782.1 | - | 1 | 1868 | |||
BC022033.2 | - | 1 | 1951 | 4825536 | ||
CR533558.1 | - | 1 | 1642 | |||
FJ150424.1 | - | 1 | 1951 |