Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145313.2 | NP_001138785.1 | 5 | 615 | ||
NM_001287191.1 | NP_001274120.1 | 4 | 519 | |||
NM_001287192.1 | NP_001274121.1 | 4 | 519 | |||
NM_031919.4 | NP_114125.1 | 4 | 519 | |||
XM_005252254.2 | XP_005252311.1 | 5 | 848 | |||
XM_005252257.3 | XP_005252314.1 | 4 | 514 | |||
XM_005252260.3 | XP_005252317.1 | 4 | 504 | |||
XM_011519077.2 | XP_011517379.1 | 6 | 506 | |||
XM_011519078.2 | XP_011517380.2 | 5 | 513 | |||
XM_011519079.2 | XP_011517381.2 | 5 | 515 | |||
XM_017015182.1 | XP_016870671.1 | 5 | 848 | |||
XM_017015183.1 | XP_016870672.1 | 4 | 514 | |||
XM_017015185.1 | XP_016870674.1 | 5 | 521 | |||
XM_017015187.1 | XP_016870676.1 | 4 | 514 | |||
GenBank mRNA | AF088031.1 | - | 4 | 508 | ||
AF316830.1 | - | 4 | 382 | |||
AK296433.1 | - | 4 | 430 | |||
AK299350.1 | - | 5 | 547 | |||
AK299491.1 | - | 4 | 430 | |||
AK309817.1 | - | 5 | 856 | |||
BP229804.1 | - | 4 | 519 | |||
BP365251.1 | - | 4 | 509 | |||
DN999402.1 | - | 4 | 362 |