Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142624.2 | NP_001136096.2 | 6 | 518 | ||
NM_001142625.2 | NP_001136097.2 | 6 | 521 | |||
NM_001144942.1 | NP_001138414.1 | 5 | 970 | |||
NM_001144943.1 | NP_001138415.1 | 6 | 518 | |||
NM_001256276.1 | NP_001243205.1 | 5 | 586 | |||
NM_001256277.1 | NP_001243206.1 | 6 | 652 | |||
NM_001256278.1 | NP_001243207.1 | 4 | 347 | |||
NM_031934.5 | NP_114140.4 | 5 | 970 | |||
GenBank mRNA | AF322067.1 | - | 5 | 346 | ||
AF327350.1 | - | 6 | 404 | |||
AJ277106.1 | - | 6 | 557 | |||
AK027312.1 | - | 6 | 551 | |||
AK074689.1 | - | 5 | 948 | |||
AK297850.1 | - | 6 | 426 | |||
AK304633.1 | - | 5 | 501 | |||
BC016841.1 | - | 5 | 828 | 3902651 | ||
BC066904.1 | - | 6 | 567 | 5260472 | ||
BC091510.1 | - | 5 | 512 | 30512024 | ||
BG749591.1 | - | 4 | 273 | |||
BT006702.1 | - | 5 | 346 | |||
CB127976.1 | - | 5 | 485 | |||
CX788239.1 | - | 6 | 411 |