Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317781.1 | NP_001304710.1 | 10 | 1678 | ||
XM_005263267.4 | XP_005263324.1 | 10 | 2121 | |||
XM_011532302.2 | XP_011530604.1 | 12 | 2256 | |||
XM_011532303.2 | XP_011530605.1 | 12 | 2253 | |||
XM_011532304.2 | XP_011530606.1 | 11 | 2220 | |||
XM_011532305.2 | XP_011530607.1 | 11 | 2157 | |||
XM_011532306.2 | XP_011530608.1 | 12 | 2256 | |||
XM_011532307.2 | XP_011530609.1 | 12 | 2256 | |||
XM_011532308.2 | XP_011530610.1 | 11 | 2220 | |||
XM_017008680.1 | XP_016864169.1 | 11 | 2220 | |||
XM_017008683.1 | XP_016864172.1 | 9 | 1165 | |||
XM_017008684.1 | XP_016864173.1 | 11 | 1437 | |||
XM_017008685.1 | XP_016864174.1 | 5 | 883 | |||
GenBank mRNA | AF113223.1 | - | 4 | 563 | ||
AK095661.1 | - | 7 | 1082 | |||
AY044656.1 | - | 3 | 478 | |||
AY601647.1 | - | 10 | 1678 | |||
BC029868.1 | - | 5 | 918 | 5171892 |