Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010984.3 | NP_001010984.1 | 4 | 1075 | ||
NM_001261462.2 | NP_001248391.1 | 3 | 946 | |||
NM_032018.6 | NP_114407.3 | 4 | 1075 | |||
XM_006711818.2 | XP_006711881.1 | 3 | 918 | |||
XM_011544289.2 | XP_011542591.1 | 2 | 401 | |||
GenBank mRNA | AK027317.1 | - | 3 | 617 | ||
AK027613.1 | - | 4 | 755 | |||
AL512744.1 | - | 4 | 689 | |||
AY358611.1 | - | 4 | 1908 | |||
BC015740.1 | - | 4 | 963 | |||
BC068478.1 | - | 4 | 1026 | 5298048 |