Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032020.4 | NP_114409.2 | 7 | 1715 | ||
GenBank mRNA | AK075458.1 | - | 7 | 1521 | ||
AY358551.1 | - | 7 | 1532 | |||
BC003060.2 | - | 7 | 1496 | 3505533 | ||
BC051268.1 | - | 7 | 1550 | 4855296 | ||
BX537633.1 | - | 3 | 2581 |