Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005149.2 | NP_005140.1 | 1 | 224 | ||
GenBank mRNA | AJ010277.1 | - | 1 | 224 | ||
BC093664.1 | - | 1 | 209 | 7939509 | ||
BC093666.1 | - | 1 | 209 | 7939511 |