Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032040.4 | NP_114429.2 | 1 | 2066 | ||
GenBank mRNA | AK297668.1 | - | 2 | 1513 | ||
AL136609.1 | - | 1 | 1992 | |||
BC025243.1 | - | 1 | 1437 | 4553965 | ||
JN703457.1 | - | 1 | 1988 | |||
KT894208.1 | - | 1 | 1222 |