Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCALD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040624.1 | NP_001035714.1 | 8 | 2057 | ||
NM_001040625.1 | NP_001035715.1 | 7 | 1970 | |||
NM_001040626.1 | NP_001035716.1 | 7 | 1970 | |||
NM_001040627.1 | NP_001035717.1 | 6 | 1883 | |||
NM_001040628.1 | NP_001035718.1 | 8 | 2026 | |||
NM_001040629.1 | NP_001035719.1 | 7 | 1939 | |||
NM_001040630.1 | NP_001035720.1 | 6 | 2123 | |||
NM_032041.2 | NP_114430.2 | 4 | 2032 | |||
XM_011517332.2 | XP_011515634.1 | 4 | 1821 | |||
XM_011517333.2 | XP_011515635.1 | 8 | 1968 | |||
XM_011517334.2 | XP_011515636.1 | 5 | 1852 | |||
XM_011517335.2 | XP_011515637.1 | 6 | 1944 | |||
XM_017013901.1 | XP_016869390.1 | 7 | 2160 | |||
XM_017013902.1 | XP_016869391.1 | 5 | 1831 | |||
GenBank mRNA | AB209015.1 | - | 5 | 4848 | ||
AF052142.1 | - | 1 | 48 | |||
AF251061.1 | - | 6 | 1808 | |||
AK055073.1 | - | 4 | 1791 | |||
AK096515.1 | - | 8 | 1982 | |||
AL136612.1 | - | 4 | 1783 | |||
BC036098.1 | - | 6 | 1808 | 5274150 |