Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014380.2 | NP_001014402.1 | 3 | 308 | ||
NM_032116.4 | NP_115492.1 | 3 | 350 | |||
XM_005266574.2 | XP_005266631.1 | 3 | 492 | |||
XM_017020791.1 | XP_016876280.1 | 3 | 363 | |||
XM_017020792.1 | XP_016876281.1 | 3 | 384 | |||
GenBank mRNA | AK097423.1 | - | 3 | 280 | ||
AK291439.1 | - | 3 | 283 | |||
AK315724.1 | - | 3 | 287 | |||
BC000612.2 | - | 3 | 310 | 3347282 | ||
DA085487.1 | - | 3 | 310 | |||
DR006891.1 | - | 3 | 385 |