Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261467.1 | NP_001248396.1 | 2 | 91 | ||
NM_032141.3 | NP_115517.1 | 3 | 185 | |||
XM_011525345.2 | XP_011523647.1 | 3 | 100 | |||
XM_017025209.1 | XP_016880698.1 | 4 | 304 | |||
XM_017025210.1 | XP_016880699.1 | 3 | 1015 | |||
XM_017025211.1 | XP_016880700.1 | 4 | 213 | |||
XM_017025212.1 | XP_016880701.1 | 3 | 350 | |||
XM_017025213.1 | XP_016880702.1 | 2 | 130 | |||
XM_017025214.1 | XP_016880703.1 | 2 | 159 | |||
XM_017025215.1 | XP_016880704.1 | 2 | 163 | |||
GenBank mRNA | AF161358.1 | - | 3 | 164 | ||
AK094552.1 | - | 1529 | ||||
AK297102.1 | - | 2 | 85 | |||
AK313963.1 | - | 3 | 172 | |||
AL136806.1 | - | 3 | 176 | |||
BC016313.1 | - | 4 | 199 | |||
BC040118.1 | - | 3 | 165 | 5176197 | ||
BC046116.2 | - | 3 | 187 | |||
BC062425.1 | - | 3 | 177 | |||
BC105044.1 | - | 3 | 172 | 8144047 | ||
BC105046.1 | - | 3 | 172 | 8144049 | ||
BC143539.1 | - | 2 | 78 | |||
CB957325.1 | - | 4 | 275 | |||
CR533555.1 | - | 3 | 148 | |||
DC327004.1 | - | 2 | 91 |