Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032142.3 | NP_115518.3 | 16 | 2493 | ||
XM_005258107.3 | XP_005258164.1 | 16 | 2548 | |||
XM_005258109.3 | XP_005258166.1 | 15 | 2338 | |||
XM_005258110.1 | XP_005258167.1 | 3 | 732 | |||
XM_006722326.3 | XP_006722389.1 | 16 | 2548 | |||
XM_006722327.3 | XP_006722390.1 | 15 | 2338 | |||
XM_006722330.3 | XP_006722393.1 | 10 | 1531 | |||
XM_011525673.2 | XP_011523975.1 | 16 | 2548 | |||
XM_011525675.2 | XP_011523977.1 | 13 | 2097 | |||
XM_017025803.1 | XP_016881292.1 | 15 | 2383 | |||
XM_017025804.1 | XP_016881293.1 | 10 | 1531 | |||
XM_017025805.1 | XP_016881294.1 | 5 | 872 | |||
XM_017025806.1 | XP_016881295.1 | 13 | 1978 | |||
XM_017025807.1 | XP_016881296.1 | 9 | 1590 | |||
GenBank mRNA | AK096548.1 | - | 13 | 1716 | ||
AL136818.1 | - | 1 | 146 | |||
AL833494.2 | - | 7 | 1211 | |||
BC144480.1 | - | 6 | 1029 | 9053007 | ||
BC144481.1 | - | 7 | 1239 | 9053008 | ||
BX648232.1 | - | 11 | 1804 | |||
BX649151.1 | - | 3 | 732 | |||
KJ567064.1 | - | 15 | 2413 |