Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM40L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286373.1 | NP_001273302.1 | 3 | 503 | ||
NM_001286374.1 | NP_001273303.1 | 3 | 503 | |||
NM_032174.5 | NP_115550.2 | 3 | 503 | |||
XM_006711572.2 | XP_006711635.1 | 3 | 378 | |||
XM_011510057.2 | XP_011508359.1 | 3 | 266 | |||
XM_017002481.1 | XP_016857970.1 | 3 | 341 | |||
XM_017002482.1 | XP_016857971.1 | 3 | 371 | |||
GenBank mRNA | AK027704.1 | - | 3 | 382 | ||
AK297848.1 | - | 3 | 382 | |||
AL523459.3 | - | 3 | 360 | |||
BC014946.1 | - | 2 | 356 | 4872309 | ||
BX537951.1 | - | 3 | 380 | |||
DA160426.1 | - | 3 | 399 | |||
DA199234.1 | - | 3 | 403 |