Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006166.3 | NP_006157.1 | 3 | 279 | ||
XM_005268908.4 | XP_005268965.1 | 4 | 343 | |||
XM_005268909.4 | XP_005268966.1 | 4 | 326 | |||
XM_005268910.4 | XP_005268967.1 | 4 | 998 | |||
GenBank mRNA | AK291934.1 | - | 3 | 211 | ||
BC005316.1 | - | 3 | 228 | 3680937 | ||
BC005317.1 | - | 3 | 234 | 3997299 | ||
BG776259.1 | - | 3 | 280 | |||
L06145.1 | - | 1 | 78 | |||
X59710.1 | - | 3 | 183 |