Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289119.1 | NP_001276048.1 | 2 | 275 | ||
NM_001289120.1 | NP_001276049.1 | 2 | 298 | |||
NM_001289121.1 | NP_001276050.1 | 2 | 416 | |||
NM_032226.2 | NP_115602.2 | 2 | 271 | |||
XM_005251608.4 | XP_005251665.1 | 2 | 1180 | |||
XM_005251612.2 | XP_005251669.1 | 2 | 238 | |||
XM_011518050.1 | XP_011516352.1 | 2 | 266 | |||
XM_017015197.1 | XP_016870686.1 | 2 | 393 | |||
XM_017015198.1 | XP_016870687.1 | 2 | 879 | |||
XM_017015199.1 | XP_016870688.1 | 2 | 240 | |||
XM_017015200.1 | XP_016870689.1 | 2 | 513 | |||
GenBank mRNA | AK026264.1 | - | 2 | 161 | ||
AK315128.1 | - | 2 | 288 | |||
BC005990.1 | - | 2 | 204 | |||
BC022434.1 | - | 3 | 364 | |||
BC036940.1 | - | 2 | 389 | 5399765 | ||
BP216150.1 | - | 2 | 298 | |||
BP246540.1 | - | 2 | 271 |