Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006193.2 | NP_006184.2 | 1, 2 | 334 | ||
XM_011516276.2 | XP_011514578.1 | 4, 5 | 608 | |||
GenBank mRNA | AB008913.1 | - | 1, 2 | 152 | ||
AF043978.1 | - | 1, 2 | 334 | |||
BC074761.2 | - | 1, 2 | 161 | 30915220 |