Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF541 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277075.1 | NP_001264004.1 | 7 | 2966 | ||
XM_005259311.4 | XP_005259368.1 | 8 | 3906 | |||
XM_005259314.3 | XP_005259371.1 | 6 | 1992 | |||
XM_011527367.2 | XP_011525669.1 | 8 | 3921 | |||
XM_011527368.2 | XP_011525670.1 | 9 | 3259 | |||
XM_011527369.2 | XP_011525671.1 | 9 | 3263 | |||
XM_011527370.2 | XP_011525672.1 | 8 | 3120 | |||
XM_011527371.2 | XP_011525673.1 | 8 | 3107 | |||
XM_011527372.2 | XP_011525674.1 | 8 | 3921 | |||
XM_011527373.2 | XP_011525675.1 | 7 | 3747 | |||
XM_011527374.2 | XP_011525676.1 | 8 | 3756 | |||
XM_017027351.1 | XP_016882840.1 | 8 | 3921 | |||
GenBank mRNA | AK314169.1 | - | 4 | 1525 | ||
AL136846.1 | - | 5 | 1335 | |||
BC101050.2 | - | 5 | 1332 | 40008070 | ||
BC101051.2 | - | 5 | 1332 | 40008071 | ||
BC101052.1 | - | 5 | 1332 | 40008072 | ||
BC101053.2 | - | 5 | 1332 | 40008073 |