Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136046.2 | NP_001129518.1 | 2 | 542 | ||
NM_001267822.1 | NP_001254751.1 | 2 | 542 | |||
NM_032265.2 | NP_115641.1 | 2 | 542 | |||
XM_017025218.1 | XP_016880707.1 | 2 | 533 | |||
GenBank mRNA | AK302181.1 | - | 2 | 533 | ||
AK310469.1 | - | 1 | 995 | |||
AL136893.1 | - | 1 | 515 |