Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM40L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286373.1 | NP_001273302.1 | 7, 8 | 910 | ||
NM_001286374.1 | NP_001273303.1 | 6, 7 | 817 | |||
NM_032174.5 | NP_115550.2 | 8, 9 | 1012 | |||
XM_006711572.2 | XP_006711635.1 | 8, 9 | 887 | |||
XM_011510057.2 | XP_011508359.1 | 6, 7 | 580 | |||
XM_017002481.1 | XP_016857970.1 | 8, 9 | 850 | |||
XM_017002482.1 | XP_016857971.1 | 7, 8 | 778 | |||
GenBank mRNA | AK022832.1 | - | 5, 6 | 628 | ||
AK027704.1 | - | 8, 9 | 891 | |||
AK297831.1 | - | 7, 8 | 664 | |||
AK297848.1 | - | 7, 8 | 789 | |||
AL523459.3 | - | 6, 7 | 674 | |||
BC014946.1 | - | 7, 8 | 865 | 4872309 | ||
BX537951.1 | - | 8, 9 | 889 |