Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135032.1 | NP_001128504.1 | 4 | 800 | ||
NM_032181.2 | NP_115557.1 | 4 | 760 | |||
GenBank mRNA | AF177337.1 | - | 834 | |||
AK023453.1 | - | 4 | 760 | |||
BC016157.1 | - | 4 | 736 | 2989786 | ||
BC063016.1 | - | 6 | 866 | 5226973 | ||
CR457314.1 | - | 2 | 322 |