Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256417.2 | NP_001243346.1 | 8 | 1296 | ||
NM_032264.4 | NP_115640.1 | 9 | 1506 | |||
NR_046176.2 | 12 | 2035 | ||||
XM_006710958.1 | XP_006711021.1 | 12 | 1820 | |||
XM_006710959.2 | XP_006711022.1 | 10 | 1604 | |||
XM_011542276.1 | XP_011540578.1 | 13 | 2077 | |||
XM_011542277.1 | XP_011540579.1 | 13 | 2062 | |||
XM_011542278.1 | XP_011540580.1 | 13 | 1891 | |||
XM_011542281.1 | XP_011540583.1 | 10 | 1427 | |||
XM_017002498.1 | XP_016857987.1 | 13 | 1876 | |||
XM_017002499.1 | XP_016857988.1 | 8 | 1142 | |||
GenBank mRNA | AK095602.1 | - | 12 | 1988 | ||
AK292580.1 | - | 9 | 1371 | |||
AK299840.1 | - | 8 | 1258 | |||
AK314597.1 | - | 9 | 1434 | |||
AL136890.1 | - | 9 | 1283 | |||
AL834330.1 | - | 5 | 479 | |||
AY598326.1 | - | 8 | 1108 | |||
AY894564.1 | - | 8 | 1012 |