Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032271.2 | NP_115647.2 | 13, 14 | 1370 | ||
XM_005255627.4 | XP_005255684.1 | 13, 14 | 1410 | |||
XM_011522700.1 | XP_011521002.1 | 13, 14 | 1412 | |||
GenBank mRNA | AK090624.1 | - | 13, 14 | 1384 | ||
AL136921.1 | - | 12, 13 | 1266 | |||
AY569455.1 | - | 12, 13 | 1323 | |||
BC024267.1 | - | 11, 12 | 1156 |