Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006593.3 | NP_006584.1 | 1 | 881 | ||
GenBank mRNA | AK226050.1 | - | 1 | 644 | ||
AK312567.1 | - | 1 | 656 | |||
BC029289.1 | - | 1 | 642 | |||
BC104844.1 | - | 1 | 809 | 8143847 | ||
BC113418.1 | - | 1 | 809 | 8322470 | ||
U49250.1 | - | 1 | 588 |