Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GARNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286779.1 | NP_001273708.1 | 7, 8 | 839 | ||
NM_032293.4 | NP_115669.3 | 6, 7 | 976 | |||
NR_104591.1 | 6, 7 | 976 | ||||
XM_005252267.3 | XP_005252324.1 | 8, 9 | 831 | |||
XM_005252268.2 | XP_005252325.1 | 8, 9 | 893 | |||
XM_011519086.2 | XP_011517388.1 | 9, 10 | 903 | |||
XM_011519087.2 | XP_011517389.1 | 9, 10 | 886 | |||
XM_011519089.2 | XP_011517391.1 | 8, 9 | 789 | |||
XM_011519090.2 | XP_011517392.1 | 7, 8 | 672 | |||
XM_017015202.1 | XP_016870691.1 | 7, 8 | 869 | |||
GenBank mRNA | AK293732.1 | - | 6, 7 | 874 | ||
AK294970.1 | - | 6, 7 | 478 | |||
AK296246.1 | - | 7, 8 | 839 | |||
AL136573.1 | - | 6, 7 | 530 | |||
BC034983.2 | - | 6, 7 | 586 | 4821143 |